Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368434.4 | NP_001355511.1 | 13 | 1438 | 97 | |
NM_066131.8 | NP_498532.4 | 12 - 13 | 1407 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |