Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368434.4 | NP_001355511.1 | 6 | 670 | 75 | |
NM_066131.8 | NP_498532.4 | 6 - 7 | 696 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |