Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025942.4 | NP_001021113.1 | 15 - 16 | 2135 | 76 | |
NM_001025943.3 | NP_001021114.1 | 17 - 18 | 2387 | 76 | ||
GenBank mRNA | AF318606.2 | - | 17 - 18 | 2260 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |