Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025942.4 | NP_001021113.1 | 9 - 10 | 1218 | 97 | |
NM_001025943.3 | NP_001021114.1 | 11 - 12 | 1470 | 97 | ||
GenBank mRNA | AF318606.2 | - | 11 - 12 | 1343 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |