Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mml-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379792.2 | NP_001367742.1 | 2 - 3 | 381 | 105 | |
NM_066230.6 | NP_498631.2 | 2 - 3 | 387 | 105 | ||
GenBank mRNA | AF264757.1 | - | 2 - 3 | 381 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |