Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mml-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379792.2 | NP_001367742.1 | 6 - 7 | 2278 | 61 | |
NM_066230.6 | NP_498631.2 | 6 - 7 | 2284 | 61 | ||
GenBank mRNA | AF264757.1 | - | 6 - 7 | 2278 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |