Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mml-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379792.2 | NP_001367742.1 | 8 - 9 | 2642 | 82 | |
NM_066230.6 | NP_498631.2 | 8 - 9 | 2648 | 82 | ||
GenBank mRNA | AF264757.1 | - | 8 - 9 | 2642 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |