Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382915.1 | 3 - 4 | 946 | 84 | ||
GenBank mRNA | AF303249.1 | - | 3 - 4 | 946 | 84 | |
L07754.1 | - | 3 - 4 | 948 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |