Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382917.1 | 16 - 17 | 4023 | 68 | ||
GenBank mRNA | AF400665.1 | - | 8 - 9 | 2353 | 68 | |
AJ320257.1 | - | 8 - 9 | 2347 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |