Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066277.5 | NP_498678.3 | 20 - 21 | 6361 | 70 | |
GenBank mRNA | AF245435.1 | - | 20 - 21 | 6338 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |