Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066277.5 | NP_498678.3 | 8 - 9 | 2333 | 95 | |
GenBank mRNA | AF245435.1 | - | 8 - 9 | 2310 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |