Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001312923.2 | NP_001299852.1 | 5 - 6 | 870 | 75 | |
NM_001312998.3 | NP_001299927.1 | 3 - 4 | 606 | 75 | ||
NM_001379801.1 | NP_001368538.1 | 4 - 5 | 768 | 75 | ||
NM_001379802.1 | NP_001367412.1 | 4 - 5 | 785 | 75 | ||
GenBank mRNA | AF047027.1 | - | 2 - 3 | 554 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |