Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066288.6 | NP_498689.2 | 3 - 4 | 386 | 79 | |
NM_181943.6 | NP_871672.2 | 3 - 4 | 364 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |