Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066346.6 | NP_498747.2 | 5 - 6 | 1445 | 90 | |
GenBank mRNA | AF104917.1 | - | 5 - 6 | 1448 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |