Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026020.7 | NP_001021191.2 | 4 - 5 | 858 | 99 | |
NM_001322687.4 | NP_001309473.1 | 3 - 4 | 650 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |