Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026020.7 | NP_001021191.2 | 1 - 2 | 96 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |