Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066412.4 | NP_498813.1 | 14 - 15 | 2862 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |