Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066412.4 | NP_498813.1 | 5 - 6 | 1143 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |