Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rfp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027528.5 | NP_001022699.1 | 1 - 2 | 124 | 149 | |
NM_001027529.6 | NP_001022700.1 | 1 - 2 | 127 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |