Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pac-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136361.5 | NP_001129833.1 | 9 - 10 | 1588 | 87 | |
NM_001306725.3 | NP_001293654.1 | 1 - 2 | 98 | 87 | ||
NM_001306727.4 | NP_001293656.1 | 3 - 4 | 558 | 87 | ||
NM_001306728.4 | NP_001293657.1 | 9 - 10 | 1598 | 87 | ||
NM_066476.7 | NP_498877.4 | 9 - 10 | 1499 | 87 | ||
GenBank mRNA | EU752496.1 | - | 9 - 10 | 1588 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |