Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382943.1 | 3 - 4 | 626 | 116 | ||
GenBank mRNA | AB120730.1 | - | 3 - 4 | 626 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |