Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382944.1 | 3 - 4 | 288 | 88 | ||
GenBank mRNA | U10327.1 | - | 2 - 3 | 97 | 88 | |
U34778.1 | - | 1 - 2 | 53 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |