Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prx-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066546.8 | NP_498947.1 | 2 - 3 | 617 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |