Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pde-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027534.6 | NP_001022705.2 | 2 - 3 | 327 | 80 | |
NM_001027535.6 | NP_001022706.2 | 1 - 2 | 231 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |