Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nac-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268047.2 | NP_001254976.1 | 10 - 11 | 1532 | 160 | |
NM_001268048.2 | NP_001254977.1 | 9 - 10 | 1279 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |