Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066620.6 | NP_499021.1 | 11 - 12 | 2840 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |