Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eat-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306696.3 | NP_001293625.1 | 9 - 10 | 1206 | 113 | |
NM_066622.6 | NP_499023.3 | 10 - 11 | 1298 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |