Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eat-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306696.3 | NP_001293625.1 | 4 - 5 | 500 | 81 | |
NM_066622.6 | NP_499023.3 | 5 - 6 | 592 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |