Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tax-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066632.6 | NP_499033.1 | 9 - 10 | 2043 | 96 | |
GenBank mRNA | D86922.1 | - | 9 - 10 | 2061 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |