Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025962.3 | NP_001021133.1 | 5 - 6 | 879 | 63 | |
NM_001025963.6 | NP_001021134.1 | 5 - 6 | 885 | 63 | ||
NM_001306716.3 | NP_001293645.1 | 2 - 3 | 563 | 63 | ||
NM_066679.5 | NP_499080.3 | 4 - 5 | 2171 | 63 | ||
GenBank mRNA | AF241656.1 | - | 5 - 6 | 932 | 63 |
Gene Symbol: B0464.9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066683.5 | NP_499084.1 | 5 - 6 | 889 | 63 |
Gene Symbol: baf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382964.1 | 2 - 3 | 549 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |