Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026054.4 | NP_001021225.1 | 12 - 13 | 3904 | 115 | |
NM_001268083.3 | NP_001255012.1 | 12 - 13 | 3963 | 115 | ||
NM_001379863.1 | NP_001366984.1 | 11 - 12 | 3801 | 115 | ||
NM_066714.4 | NP_499115.2 | 12 - 13 | 3906 | 115 | ||
GenBank mRNA | AF049142.1 | - | 12 - 13 | 3964 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |