Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026054.4 | NP_001021225.1 | 4 - 5 | 606 | 113 | |
NM_001268083.3 | NP_001255012.1 | 4 - 5 | 665 | 113 | ||
NM_001379863.1 | NP_001366984.1 | 3 - 4 | 464 | 113 | ||
NM_066714.4 | NP_499115.2 | 4 - 5 | 614 | 113 | ||
GenBank mRNA | AF049142.1 | - | 4 - 5 | 627 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |