Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026054.4 | NP_001021225.1 | 8 - 9 | 3076 | 78 | |
NM_001268083.3 | NP_001255012.1 | 8 - 9 | 3135 | 78 | ||
NM_001379863.1 | NP_001366984.1 | 7 - 8 | 2973 | 78 | ||
GenBank mRNA | AF049142.1 | - | 8 - 9 | 3136 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |