Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066733.5 | NP_499134.2 | 7 - 8 | 1688 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |