Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066733.5 | NP_499134.2 | 8 - 9 | 1783 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |