Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025987.5 | NP_001021158.1 | 2 - 3 | 309 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |