Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066812.7 | NP_499213.1 | 5 - 6 | 2166 | 90 | |
GenBank mRNA | U73679.1 | - | 5 - 6 | 2171 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |