Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027542.4 | NP_001022713.1 | 8 - 9 | 2453 | 83 | |
NM_001027544.3 | NP_001022715.1 | 5 - 6 | 1300 | 83 | ||
NM_066812.7 | NP_499213.1 | 7 - 8 | 2396 | 83 | ||
GenBank mRNA | U73679.1 | - | 7 - 8 | 2401 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |