Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mua-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027503.4 | NP_001022674.1 | 26 - 27 | 5582 | 107 | |
NM_001027504.3 | NP_001022675.1 | 16 - 17 | 4222 | 107 | ||
GenBank mRNA | AF139060.1 | - | 26 - 27 | 5582 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |