Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mua-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027503.4 | NP_001022674.1 | 32 - 33 | 10834 | 80 | |
NM_001027504.3 | NP_001022675.1 | 22 - 23 | 9474 | 80 | ||
GenBank mRNA | AF139060.1 | - | 32 - 33 | 10834 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |