Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066885.7 | NP_499286.2 | 4 - 5 | 606 | 84 | |
GenBank mRNA | AB019521.1 | - | 2 - 3 | 398 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |