Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268143.3 | NP_001255072.1 | 11 - 12 | 2023 | 71 | |
NM_001268144.3 | NP_001255073.1 | 7 - 8 | 1224 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |