Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027611.3 | NP_001022782.1 | 10 - 11 | 1325 | 85 | |
NM_001379903.1 | NP_001368253.1 | 6 - 7 | 698 | 85 | ||
GenBank mRNA | AF151640.1 | - | 10 - 11 | 1494 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |