Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027615.4 | NP_001022786.1 | 5 - 6 | 769 | 85 | |
NM_001306734.3 | NP_001293663.1 | 1 - 2 | 59 | 85 | ||
GenBank mRNA | AF151644.1 | - | 5 - 6 | 676 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |