Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hpr-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379907.1 | NP_001368495.1 | 5 - 6 | 460 | 118 | |
NM_066941.7 | NP_499342.3 | 5 - 6 | 538 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |