Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnt-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027665.3 | NP_001022836.1 | 3 - 4 | 1011 | 89 | |
NM_001027666.5 | NP_001022837.1 | 5 - 6 | 435 | 89 | ||
NM_001027667.5 | NP_001022838.1 | 2 - 3 | 254 | 89 | ||
NM_001382987.1 | 4 - 5 | 372 | 89 | |||
NM_001382988.1 | 5 - 6 | 432 | 89 | |||
NM_001382989.1 | 5 - 6 | 411 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |