Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hpl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027481.5 | NP_001022652.1 | 3 - 4 | 465 | 88 | |
GenBank mRNA | AF123574.1 | - | 3 - 4 | 465 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Caenorhabditis elegans Probe spans exons |