Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klp-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027506.4 | NP_001022677.1 | 5 - 6 | 1628 | 62 | |
NM_001027507.4 | NP_001022678.1 | 4 - 5 | 1454 | 62 | ||
GenBank mRNA | AB017164.1 | - | 4 - 5 | 1460 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |