Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: athp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066996.6 | NP_499397.2 | 9 - 10 | 2268 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |