Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vab-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067000.5 | NP_499401.1 | 4 - 5 | 581 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |